Anti-Aquaporin 4 Rabbit Polyclonal Antibody

Proveedor: Boster
BSBTPB9475EA 550 EUR
BSBTPB9475
Anti-Aquaporin 4 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for Aquaporin-4(AQP4) detection. Tested with WB, IHC-P, IHC-F in Human;Mouse;Rat.

Type: Primary
Antigen: Aquaporin 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR