Anti-Aquaporin 2 Rabbit Polyclonal Antibody

Proveedor: Boster
BSBTPB9474EA 550 EUR
BSBTPB9474
Anti-Aquaporin 2 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for Aquaporin-2(AQP2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: Aquaporin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR