Anti-AGTR1 Rabbit Polyclonal Antibody

Proveedor: Boster
BSBTPB9470EA 550 EUR
BSBTPB9470
Anti-AGTR1 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for Type-1 angiotensin II receptor(AGTR1) detection. Tested with WB, IHC-P in Human;Mouse.

Type: Primary
Antigen: AGTR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR