Hybridoma medium, UltraDOMA™
Proveedor: Lonza
733-1759EA 0 EUR
733-1759 733-1762
Hybridoma medium, UltraDOMA™
Medios de cultivo celular
UltraDOMA™ Serum-free Hybridoma Medium is a formulation designed for the cultivation of murine, human, and chimeric hybridomas in batch culture and in hollow fibre bioreactors. UltraDOMA™ Medium is supplemented with recombinant human insulin, bovine transferrin and bovine albumin. The total protein concentration is 30 μg/ml. UltraDOMA ™ Medium does not contain L-glutamine.
UltraDOMA-PF™ is a protein-free formulation designed for the cultivation of murine, human and chimeric hybridomas. The lack of proteins in the formulation speeds downstream processing and purification. UltraDOMA-PF™ contains L-glutamine.
UltraDOMA™ Medium contains no human-derived proteins.