Anti-THY12 Rabbit Polyclonal Antibody

Proveedor: AbFrontier
ABFRLF-PA0227EA 295 EUR
ABFRLF-PA0227
Anti-THY12 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: THY12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR