Anti-the Tyrosine Kinase Receptor C ECD Rabbit Polyclonal Antibody

Proveedor: Biosensis
BSENR-151-100EA 302 EUR
BSENR-151-100
Anti-the Tyrosine Kinase Receptor C ECD Rabbit Polyclonal Antibody
Anticuerpos
TrkC is a member of the neurotrophic tyrosine receptor kinase family. TrkC is a membrane-bound receptor that upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. TrkC is the receptor for neurotrophin-3 (NT-3).  Signalling through TrkC leads to cell differentiation and may play a role in the development of proprioceptive neurons that sense body position. SUBUNIT: Exists in a dynamic equilibrium between monomeric (low affinity) and dimeric (high affinity) structures. SUBCELLULAR LOCATION: Membrane; single-pass type I membrane protein. ALTERNATIVE PRODUCTS: 4 named isoforms produced by alternative splicing. Additional isoforms seem to exist. Mutations in TrkC have been associated with medulloblastomas, secretory breast carcinomas and other cancers.

Specificity was demonstrated by immunohistochemistry. This antibody was used to stain cryostat sections of the rat peripheral sensory ganglia.
Application Information:
IHC. A dilution of 1:1000 to 1:3000 is recommended for this application. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: the Tyrosine Kinase Receptor C ECD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple epitopes because of the nature of the immunogen
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR