Neurotensin is a neuropeptide which regulates secretion of luteinizing hormone and prolactin. Involved in dopamine neurotransmission.
Antibody recognizes Neurotensin by native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Cross reactivity: Human; mouse; rat. Highly conserved and expected to interact with bovine, feline and monkey
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Neurotensin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Neurotensin peptide (151-163 aa)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat