Anti-Leptin Rabbit Polyclonal Antibody

Proveedor: Biosensis
R-1552-100
BSENR-1552-100EA 312 EUR
BSENR-1552-100
Anti-Leptin Rabbit Polyclonal Antibody
Anticuerpos
Leptin is secreted by white adipocytes and functions as part of a signaling pathway that can inhibit food intake and/or regulate energy expenditure to maintain constancy of the adipose mass. Leptin has a variety of other roles including endocrine functions, regulation of immune and inflammatory responses, hematopoiesis, angiogenesis and wound healing (Ref Entrez Gene).

Antibody recognizes Leptin in humans, mouse and rat. Approved applications include WB and ELISA. Suggested dilution of 1:1,000-1:4,000. Biosensis recommends that the optimal working dilution should be determined by the end user. Other applications not yet tested.
Application Information:
WB and ELISA. Suggested dilution of 1:1,000-1:2,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Leptin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR