Anti-AP-1 complex subunit beta-1 Rabbit Polyclonal Antibody
Proveedor: Biosensis
BSENR-167-100EA 312 EUR
BSENR-167-100
Anti-AP-1 complex subunit beta-1 Rabbit Polyclonal Antibody
Anticuerpos
FUNCTION: Subunit of clathrin-associated adaptor protein complex 1 that plays a role in protein sorting in the late-Golgi/trans-Golgi network (TGN) and/or endosomes. The AP complexes mediate both the recruitment of clathrin to membranes and the recognition of sorting signals within the cytosolic tails of transmembrane cargo molecules. SUBUNIT: Adaptor protein complex 1 (AP-1) is an heterotetramer composed of two large adaptins (gamma1/AP1G1 or gamma2/AP1G2 and beta1A/AP1B1 or beta1B/AP1B1), a medium adaptin (mu1A/AP1M1 or mu1B/AP1M2) and a small adaptin (sigma1A/AP1S1 or sigma1B/AP1S2 or sigma1C/AP1S3). SUBCELLULAR LOCATION: Golgi apparatus. Cytoplasmic vesicle; cytoplasmic vesicle membrane; peripheral membrane protein; cytoplasmic side. Note=Component of the coat surrounding the cytoplasmic face of coated vesicles located at the Golgi complex. ALTERNATIVE PRODUCTS: 2 named isoforms produced by alternative splicing. TISSUE SPECIFICITY: Widely expressed. DISEASE: Deletion of the AP1B1 gene may play a role in the tumorigenesis of meningiomas. SIMILARITY: Belongs to the adaptor complexes large subunit family.
This antibody has been shown to be specific for AP-1 complex subunit beta-1.
Application Information:
IHC, WB. This antibody works superbly in immunohistochemistry on frozen or paraffin embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for routine immunohistochemistry are 1: 1000 to 1: 5000 depending on tissue and detection method. For western blotting a dilution range of 1: 100 to 1: 10000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: AP-1 complex subunit beta-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 11-29 of Human AP-1
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human