Anti-superoxide dismutase Rabbit Polyclonal Antibody

Proveedor: Biosensis
BSENR-168-100EA 312 EUR
BSENR-168-100
Anti-superoxide dismutase Rabbit Polyclonal Antibody
Anticuerpos
SOD1 binds copper and zinc ions ans is one of two isozymes responsible for destroying free superoxide radicals which are normally produced within the cells and which are toxic to biological systems. SOD1 is a soluble cytoplasmic protein, acting as a homodimer to convert superoxide radicals to molecular oxygen and hydrogen peroxide. Defects in SOD1 are the cause of amyotrophic lateral sclerosis type 1 (ALS1) which is a neurodegenerative disorder affecting upper and lower motor neurons and resulting in fatal paralysis.

This antibody has been shown to be specific for superoxide dismutase (SOD1) protein.
Application Information:
IHC, WB. This antibody works superbly in immunohistochemistry on frozen or paraffin embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for routine immunohistochemistry are 1: 100 to 1: 1000 depending on tissue and detection method. For western blotting a dilution range of 1: 1000 to 1: 4000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: superoxide dismutase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 121-139 of Human superoxide dismutase (SOD1)
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR