Anti-Leptin, Rat Chicken Polyclonal Antibody

Proveedor: Biosensis
BSENC-1515-500EA 312 EUR
BSENC-1515-500
Anti-Leptin, Rat Chicken Polyclonal Antibody
Anticuerpos
Leptin is secreted by white adipocytes and functions as part of a signaling pathway that can inhibit food intake and/or regulate energy expenditure to maintain constancy of the adipose mass. Leptin also has several endocrine functions and is involved in the regulation of immune and inflammatory responses, hematopoiesis, angiogenesis and wound healing (Ref: Entrez Gene) .

The antibody reacts with rodent leptin, reactivity to human leptin appears minimal but has not be exhaustively tested.
Application Information:
ELISA and WB. Suggested dilution of 1:1,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Leptin, Rat
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Chicken
Isotype: IgY
Reactivity: Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR