Anti-C-reactive Protein Goat Polyclonal Antibody

Proveedor: Biosensis
BSENG-007-100EA 312 EUR
BSENG-007-100
Anti-C-reactive Protein Goat Polyclonal Antibody
Anticuerpos
Can interact with DNA and histones and may scavenge nuclear material released from damaged circulating cells. COFACTOR: Binds 2 calcium ions per subunit.

It has been shown to be specific for C-reactive protein by WB.
Application Information:
WB. Extremely high titre antibody. For western blot a dilution of 1: 10,000 to 1: 100000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: C-reactive Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR