Anti-Peptide YY Chicken Polyclonal Antibody

Proveedor: Biosensis
BSENC-1537-100EA 322 EUR
BSENC-1537-100
Anti-Peptide YY Chicken Polyclonal Antibody
Anticuerpos
Peptide YY (PYY) is secreted from endocrine cells in the lower small intestine, colon and pancreas. PYY inhibits exocrine pancreatic secretion, has a vasoconstrictory action and inhibitis jejunal and colonic mobility (Ref: SWISS-Prot).

Antibody recognizes Peptide YY in ELISAs, other applications have not been tested. This format is biotinylated and can be used as a detector antibody in ELISAs when paired with C-1538
Application Information:
ELISA . Suggested dilution at 1:2,000 to 1:10,000 (detection antibody). Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Peptide YY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminus of Human mature Peptide YY (24-36 aa) .
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR