Presenilin-2 (PSEN2) is a multi-pass membrane protein and component of the gamma-secretase complex. Defects in PSEN2 are a cause of Alzheimer disease type 4 (AD4), an autosomal dominant Alzheimer disease. (Ref:SWISS-Prot).
Antibody recognizes Presenillin-2 in western blot and by ELISA, other applications have not yet been tested.
Application Information:
WB and ELISA. Suggested dilution of 1:1,000 to 1:4,000. To minimise background staining, a higher concentration of detergent (such as Tween-20) is suggested in the dilution and washing steps. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Presenilin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple epitopes because of the nature of the immunogen
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat