Anti-PAFAH subunit gamma Mouse Monoclonal Antibody [clone: 8C11]

Proveedor: Biosensis
BSENM-856-100EA 327 EUR
BSENM-856-100
Anti-PAFAH subunit gamma Mouse Monoclonal Antibody [clone: 8C11]
Anticuerpos
Cytosolic PAFAH IB is formed of three subunits, alpha (45kDa), beta (30kDa, and gamma (29kDa). Its catalytic activity of PAFAH IB resides in the beta and gamma subunits while the alpha subunit has regulatory activity. Trimer formation is not essential for the catalytic activity. Specifically, PAFAH subunit gamma inactivates platelet-activating factor by removing the acetyl group at the sn-2 position. It plays an important role during the development of the brain. In the adult, it is expressed in brain, skeletal muscle, kidney, thymus, spleen, colon, testis, ovary and peripheral blood leukocytes. In the fetus, highest expression occurs in the brain.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, sandwich ELISA and immunofluorescence. The recommended dilution for this antibody is 25 µg/ml for immunofluorescence. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PAFAH subunit gamma
Clonality: Monoclonal
Clone: 8C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR