Neurogenin-1 appears to mediate neuronal differentiation. It requires dimerisation with another basic-helix-loop-helix protein for efficient DNA binding. Its expression is restricted to the embryonic nervous system.
Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, sandwich ELISA and immunofluorescence. The recommended dilution for this antibody is 50 µg/ml for immunofluorescence. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Neurogenin-1
Clonality: Monoclonal
Clone: 4A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human