Anti-Deafness dystonia Protein 1 Mouse Monoclonal Antibody [clone: 2F11]

Proveedor: Biosensis
BSENM-831-100EA 327 EUR
BSENM-831-100
Anti-Deafness dystonia Protein 1 Mouse Monoclonal Antibody [clone: 2F11]
Anticuerpos
TIMM8A is a mitochondrial intermembrane chaperone that is involved in the import and insertion of some multi-pass transmembrane proteins into the mitochondrial inner membrane. It also acts as a chaperone-like protein that protects the hydrophobic precursors from aggregation and guide them through the mitochondrial intermembrane space. TIMM8A is probably necessary for normal neurologic development. TIMM8A is a heterohexamer composed of three TIMM8A and three TIMM13 subunits, named soluble 70KDa complex. It associates with the TIMM22 complex. It is located at the mitochondrion inner membrane and is highly expressed in fetal and adult brain, followed by fetal lung, liver and kidney. Defects in TIMM8A cause Mohr-Tranebjaerg syndrome (MTS) also known as dystonia-deafness syndrome (DDS) or X-linked progressive deafnes type 1 (DFN-1).  This is a recessive neurodegenerative syndrome characterised by progressive deafness, dystonia, spasticity, dysphagia, mental deterioration, paranoia and cortical blindness. Defects in TIMM8A are also the cause of Jensen syndrom, also known as opticoacoustic nerve atrophy with dementia. This is an X-linked disease characterised by deafness, blindess and muscle weakness.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, IHC and sandwich ELISA. For IHC, a concentration of 3 µg/ml is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Deafness dystonia Protein 1
Clonality: Monoclonal
Clone: 2F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR