Anti-Ephrin-A51 Mouse Monoclonal Antibody [clone: 1F12]

Proveedor: Biosensis
BSENM-832-100EA 327 EUR
BSENM-832-100
Anti-Ephrin-A51 Mouse Monoclonal Antibody [clone: 1F12]
Anticuerpos
Ephrin-A5 is a member of the ephrin (EPH) gene family. The EPH and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, particularly in the nervous system. EPH receptors typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. Ephrin-A5  prevents axon bundling in cocultures of cortical neurons with astrocytes, a model of late stage nervous system development and differentiation.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Ephrin-A51
Clonality: Monoclonal
Clone: 1F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR