Anti-Orexin A Chicken Polyclonal Antibody

Proveedor: Biosensis
BSENC-1531-100EA 345 EUR
BSENC-1531-100
Anti-Orexin A Chicken Polyclonal Antibody
Anticuerpos
FUNCTION: Neuropeptides that play a significant role in the regulation of food intake and sleep-wakefulness, possibly by coordinating the complex behavioral and physiologic responses of these complementary homeostatic functions. A broader role in the homeostatic regulation of energy metabolism, autonomic function, hormonal balance and the regulation of body fluids, is also suggested. Orexin-A binds to both OX1R and OX2R with a high affinity, whereas orexin-B binds only to OX2R with a similar high affinity. SUBCELLULAR LOCATION: Endoplasmic reticulum; rough endoplasmic reticulum. Associated with perikaryal rough endoplasmic reticulum as well as cytoplasmic large granular vesicles at synapses. SIMILARITY: Belongs to the orexin family.

The antibody recognizes Orexin-A in ELISA other applications have not been tested.
Application Information:
ELISA . Suggested dilution at 1:2,000 to 1:5,000 as a detection antibody and 1:200 to 1:1,000 as a coating antibody in sandwich ELISA. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Orexin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Orexin A (1-13 aa) .
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR