Anti-Stathmin-2 Mouse Monoclonal Antibody [clone: 1C6]

Proveedor: Biosensis
BSENM-890-100EA 327 EUR
BSENM-890-100
Anti-Stathmin-2 Mouse Monoclonal Antibody [clone: 1C6]
Anticuerpos
Stathmin-2 is a neuronal growth-associated protein that shares significant amino acid sequence similarity with the phosphoprotein stathmin. It may play a role in neuronal differentiation, and in modulating membrane interaction with the cytoskeleton during neurite outgrowth. It is neuron specific and belongs to the stathmin family.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Stathmin-2
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR