Anti-ATG12 Mouse Monoclonal Antibody [clone: 2H8]

Proveedor: Biosensis
BSENM-879-100EA 327 EUR
BSENM-879-100
Anti-ATG12 Mouse Monoclonal Antibody [clone: 2H8]
Anticuerpos
Autophagy is a process of bulk protein degradation in which cytoplasmic components, including organelles, are enclosed in double-membrane structures called autophagosomes and delivered to lysosomes or vacuoles for degradation. ATG12 is the human homolog of a yeast protein involved in autophagy.  ATG12 is conjugated to ATG5. The ATG12-ATG5 conjugate has an apparent molecular mass of 65 kDa. The C-terminal glycine of ATG12 is conjugated to a central lysine (lys130) of ATG5. Studies showed that over expression of ATG3 facilitated formation of the ATG12-ATG5 conjugate, suggesting that ATG3 cross-talks with the ATG12 conjugation system. ATG12 is ubiquitously expressed.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ATG12
Clonality: Monoclonal
Clone: 2H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR