Anti-Neuropilin-1 Mouse Monoclonal Antibody [clone: 1B3]

Proveedor: Biosensis
BSENM-878-100EA 327 EUR
BSENM-878-100
Anti-Neuropilin-1 Mouse Monoclonal Antibody [clone: 1B3]
Anticuerpos
The antibody is made against the membrane-bound isoform 1 of Neuropilin-1. It is a receptor involved in the development of the cardiovascular systerm, in angiogenesis, in the formation of certain neuronal circuits and in organogenesis outside the nervous system. It binds both vascular endothelial growth factor (VEGF) and semaphorin family members. Isoform 1 is expressed by blood vessels of different tissues. In the developing embryo it is predominantly found in the nervous system. In adult tissues, it is highly expressed in heart and placenta, moderately expressed in lung, liver and skeletal muscle, kidney and pancreas, and low in adult brain.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Neuropilin-1
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR