Anti-Pleiotrophin Mouse Monoclonal Antibody [clone: 5C3]

Proveedor: Biosensis
BSENM-860-100EA 327 EUR
BSENM-860-100
Anti-Pleiotrophin Mouse Monoclonal Antibody [clone: 5C3]
Anticuerpos
Pleiotrophin is an 18 kDa growth factor that has high affinity for heparin. It also has neurite extension activity. It is a secreted protein that is expressed in osteoblasts and brain.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, IHC and sandwich ELISA. The recommended dilution for this antibody is 3 µg/ml for IHC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Pleiotrophin
Clonality: Monoclonal
Clone: 5C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR