Anti-Fucosyltransferase 10 Mouse Monoclonal Antibody [clone: 4H3]

Proveedor: Biosensis
BSENM-910-100EA 327 EUR
BSENM-910-100
Anti-Fucosyltransferase 10 Mouse Monoclonal Antibody [clone: 4H3]
Anticuerpos
Fucosyltransferase 10 is a probable fucosyltransferase that localises to the golgi apparatus, the Golgi stack membrane. It is a single-pass type II membrane protein. There are 7 named isoforms produced by alternative splicing.

Specificity has been confirmed by WB and ELISA against the antigen.
Application Information:
This antibody is recommended for WB and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Fucosyltransferase 10
Clonality: Monoclonal
Clone: 4H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR