Anti-ATG3 Mouse Monoclonal Antibody [clone: 1F7]

Proveedor: Biosensis
BSENM-908-100EA 327 EUR
BSENM-908-100
Anti-ATG3 Mouse Monoclonal Antibody [clone: 1F7]
Anticuerpos
Autophagy is a process of bulk degradation of cytoplasmic components by the lysosome or vacuole. ATG3 is an E2-like protein-conjugating enzyme for GABARAPL1, GABARAPL2 and MAPLC3 binding them to PE. ATG3 forms an E1-E2 complex with ATG7 and cross-talks with the ATG12 conjugation system. ATG3 is a cytoplasmic protein that is widely expressed with highest expression in heart, skeletal muscle, kidney , liver and placenta.

Specificity has been confirmed by WB and ELISA against the antigen.
Application Information:
This antibody is recommended for WB and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ATG3
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR