Anti-Kallikrein-8 Mouse Monoclonal Antibody [clone: 2F11]

Proveedor: Biosensis
BSENM-891-100EA 327 EUR
BSENM-891-100
Anti-Kallikrein-8 Mouse Monoclonal Antibody [clone: 2F11]
Anticuerpos
Kalllikrein-8 is a serine protease capable of degrading several proteins such as casein, fibrinogen, kininogen, fibronectin and callagen type IV. It also cleaves L1CAM in response to increased neural activity. It induces neurite outgrowth and fasciculation of cultured hippocampal neurons. It plays a role in the formation and maturation of orphan and small synaptic boutons in the Schaffer-collateral pathway, regulates Schaffer-collateral long-term potentiation in the hippocampus and is required for memory acquisition and synaptic plasticity. It is  involved in skin desquamation and keratinocyte proliferation. Plays a role in the secondary phase of pathogenesis following spinal cord injury. It is Inhibited by a range of serine protease inhibitors including antipain, aprotinin, leupeptin, benzamidine and soybean trypsin inhibitor. It is a secreted protein that has 4 named isoforms produced by alternative splicing. Isoform 1 is predominantly expressed in the pancreas while isoform 2 is expressed in adult brain and hippocampus. Both forms are also found in fetal brain and placenta. Displays an 11.5-fold increase in Alzheimer disease hippocampus compared to controls and is overexpressed in some ovarian carcinomas. Expressed at low levels in normal skin while high levels are found in psoriasis vulgaris, seborrheic keratosis, lichen planus and squamous cell carcinoma skin samples.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, immunofluorescence and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Kallikrein-8
Clonality: Monoclonal
Clone: 2F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR