Anti-VCX-A Mouse Monoclonal Antibody [clone: 6A3]

Proveedor: Biosensis
BSENM-899-100EA 327 EUR
BSENM-899-100
Anti-VCX-A Mouse Monoclonal Antibody [clone: 6A3]
Anticuerpos
VCX-A is only expressed in testis and may play a role in spermatogenesis or in sex ratio distortion. It is a 186 amino acid peptide that has an estimated molecular weight of ~20 kDa. It was recently identified that VCX-A is an inhibitor of the decapping enzyme DCP2 and that it stabilised mRNA. It is a noncanonical cap-binding protein that binds to capped RNA but not cap structures lacking RNA.  Defects in VCX-A are associated with some forms of X-linked nonspecific mental retardation.

Specificity has been confirmed by WB and ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: VCX-A
Clonality: Monoclonal
Clone: 6A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR