Anti-Protocadherin alpha 1 Mouse Monoclonal Antibody [clone: 3A3]

Proveedor: Biosensis
BSENM-907-100EA 327 EUR
BSENM-907-100
Anti-Protocadherin alpha 1 Mouse Monoclonal Antibody [clone: 3A3]
Anticuerpos
Protocadherin alpha-1 is 1 of 15 tandemly arranged genes within the PCDHA gene cluster. These 15 genes function as 'variable' exons that are individually spliced to a downstream constant region to form distinct PCDHA transcripts. They encode cadherin-like cell surface proteins that are expressed in neurons and are present at synaptic junctions.

Specificity has been confirmed by WB and ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Protocadherin alpha 1
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR