Anti-MSGA Mouse Monoclonal Antibody [clone: 7A11]

Proveedor: Biosensis
BSENM-820-100EA 327 EUR
BSENM-820-100
Anti-MSGA Mouse Monoclonal Antibody [clone: 7A11]
Anticuerpos
MSGA belongs to the intercrine alpha (chemokine CXC) familly. In this family, the first two of the four conserved residues are separated by a single amino acid. MSGA has chemotactic activity for neutrophils and may play a role in inflammation. MSA is secreted and subsequently proteolytically processed at the N-terminus by peripheral blood monocytes to form GRO-alpha(4-73), GRO-alpah(5-73) and GRO-alpha(6-73). In vitro, these processed forms show a 30 fold increase in chemotactic activity. The unprocessed precursor has a length of 107 amino acids and a predicted molecular weight of 11.3KDa.

The specificity of this antibody has been confirmed by WB and ELISA against the GST tagged recombinant human MSGA protein.
Application Information:
This antibody is recommended for WB and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MSGA
Clonality: Monoclonal
Clone: 7A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR