Catenin delta-2 functions as a transcriptional activator when bound to ZBTB33. It may also be involved in neuronal cell adhesion and tissue morphogenesis and integrity by regulating adhesion molecules. It is predominantly expressed in the brain and localises to the nucleus and cell junction. The unprocessed precursor has a length of 1225 amino acids and a predicted molecular weight of 132.7KDa. This protein is O-glycosylated. It is also phosporylated upon DNA damage. It belongs to the beta-catenin family and contains nine ARM repeats.
The specificity of this antibody has been confirmed by WB and ELISA against the antigen.
Application Information:
This antibody is recommended for WB and IHC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: catenin delta-2
Clonality: Monoclonal
Clone: 6E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse, Rat