Anti-PhyH Mouse Monoclonal Antibody [clone: 1F2-5B9]

BSENM-914-100EA 327 EUR
BSENM-914-100
Anti-PhyH Mouse Monoclonal Antibody [clone: 1F2-5B9]
Anticuerpos
PhyH is a peroxisomal protein that is involved in the alpha-oxidation of 3-methyl branched fatty acids. Specifically, this protein converts phytanoyl-CoA to 2-hydroxyphytanoyl-CoA. It requires iron and ascorbate as cofactors and is expressed in liver, kidney, and T-cells but not in spleen, brain, heart, lung and skeletal muscle. Defects in PhyH are a cause of Refsum disease (RD) and deficient protein activity has been associated with Zellweger syndrome and rhizomelic chondrodysplasia punctata. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Specificity has been confirmed by WB and ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PhyH
Clonality: Monoclonal
Clone: 1F2-5B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR