Anti-Mitochondrial Mn superoxide dismutase (SOD2) Rabbit Polyclonal Antibody

Proveedor: Biosensis
BSENR-1596-100EA 345 EUR
BSENR-1596-100
Anti-Mitochondrial Mn superoxide dismutase (SOD2) Rabbit Polyclonal Antibody
Anticuerpos
Superoxide dismutase [Mn], mitochondrial destroys superoxide anion radicals which are normally produced within the cells and which are toxic to biological systems (Ref: SwissProt).

Antibody recognizes Mn SOD by multiple applications including IF, WB. Typical working dilutions for light microscopy are 1:500 to 1:1,000 depending on tissue and detection method. For IF, a dilution range of 1:50 to 1:100 is recommended. For WB, a dilution range of 1: 1,000 to 1: 4,000 is recommended. This antibody clearly detects a protein at approximately 24 kDa on WB of human brain tissue. Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Cross reactivity is mouse, rat, and human at least.
Application Information:
IF, WB. Typical working dilutions for light microscopy are 1:500 to 1:1,000 depending on tissue and detection method. For IF, a dilution range of 1:50 to 1:100 is recommended. For WB, a dilution range of 1: 1,000 to 1: 4,000 is recommended. This antibody clearly detects a protein at approximately 24 kDa on WB of human brain tissue. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Mitochondrial Mn superoxide dismutase (SOD2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: multiple epitopes because of the nature of the immunogen
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR