Anti-Nuclear receptor TLX Mouse Monoclonal Antibody [clone: 1C4]

Proveedor: Biosensis
BSENM-920-100EA 327 EUR
BSENM-920-100
Anti-Nuclear receptor TLX Mouse Monoclonal Antibody [clone: 1C4]
Anticuerpos
Nuclear receptor TLX is an orphan receptor that binds DNA as a monomer and shares a target gene specificity that is unique among the nuclear receptor superfamily. It is thought to be required for brain development and also involved in the regulation of retinal development. It is a brain-specific protein that is present in all brain section tested with highest levels in the caudate nucleus and hippocampus.

Specificity has been confirmed by direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Nuclear receptor TLX
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR