- Pagina principal
- Anticuerpos
- Anti-p75NTR Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MC192]
Anti-p75NTR Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MC192]
Proveedor: Biosensis
BSENM-016-100-FTEA
334
EUR
BSENM-016-100-FT
Anti-p75NTR Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MC192]
Anticuerpos
Monoclonal antibody MC192 against the rat low affinity nerve growth factor receptor (p75NTR) is derived from the fusion of Sp2/0-Ag 14 myeloma cells with mouse immune splenocytes. MC192 monoclonal antibody was originally generated by Chandlers et al. p75NTR was originally discovered as a low affinity nerve growth factor receptor. Later it was found that it was the receptor for all neurotrophins. It mediates signals of neurotrophins for neuronal survival, apoptosis, neurite outgrowth and synaptic plasticity. Recently, it has been revealed that p75NTR not only acts as the receptor for neurotrophins but also the receptor for many other pathological ligands such as prions, rabies virus and amyloid beta. p75NTR also acts as a co-receptor for NOGO which mediates inhibitory signals of myelin associated protein. p75NTR is highly expressed in a number of non-neuronal and neuronal cells including motor neurons during development and also in damaged neurons. MC192 has a potential use as the ligand for gene delivery into p75NTR-expressing rat cells via a receptor-mediated mechanism. FUNCTION: Low affinity receptor which can bind to NGF, BDNF, NT-3, and NT-4. Can mediate cell survival as well as cell death of neural cells. SUBUNIT: Homodimer; disulfide-linked. Interacts with p75NTR-associated cell death executor. Interacts with NGFRAP1/BEX3. Interacts with TRAF2, TRAF4, TRAF6, PTPN13 and RANBP9.
MC192 recognizes the extracellular domain of the neurotrophin receptor p75NTR in rat. MC192 antibody may be used for immunocytochemical localisation of rat cells expressing p75NTR, ELISA and western blot. This antibody has also been used for the construction of the MC192-saporin immunotoxin for specific elimination of neuronal populations in basal forebrain cholinergic neurons to generate an animal model for Alzheimer's disease. Using Flow Cytometry, this antibody has frequently been employed for panning to isolate p75NTR-expressing rat cells.
Application Information:
Immunohistochemistry, immunofluorescence, flow cytometry, NGF receptor p75 dynamics, retrograde transport studies, study of intracellular trafficking. Suggested working dilutions: For immunohistochemistry a concentration of 1-2 Â µg/ml is recommended. The antibody is not appropriate for Western Blots . The recommended concentration for FACS is 20 Â µg/ml and at least 1 in 5000 dilution is recommended for 1 site ELISA. Optimal working dilution should be determined by the end user. MC192 is not suitable as a blocking agent, although it has been incorrectly used for this purpose in many published works. The antibody was generated specifically by screening for monoclonals that had the ability to ENHANCE the binding of NGF, the natural ligand for p75. Therefore, this antibody is particularly unusual. The full details can be found in the original paper, which is listed on our datasheet (see Chandler et al, 1984).. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Application Information:
Immunohistochemistry, immunofluorescence, flow cytometry, NGF receptor p75 dynamics, retrograde transport studies, study of intracellular trafficking. Suggested working dilutions: For immunohistochemistry a concentration of 1-2 Â µg/ml is recommended. The antibody is not appropriate for Western Blots . The recommended concentration for FACS is 20 Â µg/ml and at least 1 in 5000 dilution is recommended for 1 site ELISA. Optimal working dilution should be determined by the end user. MC192 is not suitable as a blocking agent, although it has been incorrectly used for this purpose in many published works. The antibody was generated specifically by screening for monoclonals that had the ability to ENHANCE the binding of NGF, the natural ligand for p75. Therefore, this antibody is particularly unusual. The full details can be found in the original paper, which is listed on our datasheet (see Chandler et al, 1984).. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: p75NTR
Clonality: Monoclonal
Clone: MC192
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Human Small EDRK-rich factor 1 (1-82)
Host: Mouse
Isotype:
Reactivity: Rat
Antigen: p75NTR
Clonality: Monoclonal
Clone: MC192
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Human Small EDRK-rich factor 1 (1-82)
Host: Mouse
Isotype:
Reactivity: Rat
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Consulta de precio
Learn more
About VWR
Item(s) Added to Cart