Anti-oxytocin Guinea Pig Polyclonal Antibody

Proveedor: Biosensis
BSENGP-037-50EA 357 EUR
BSENGP-037-50
Anti-oxytocin Guinea Pig Polyclonal Antibody
Anticuerpos
Oxytocin causes contraction of the smooth muscle of the uterus and of the mammary gland. It belongs to the vasopressin/oxytocin family. Oxytocin is secreted.

Specificity was demonstrated by immunohistochemistry.
Application Information:
IHC. A concentration of 1 in 2000 is recommended for IHC. IHC performed in sheep brain (hypothalamus) demonstrates intense staining of cells and terminals. No staining is evident when the primary antibody is pre-absorbed with 0.5 mg/ml of oxytocin. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: oxytocin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 20-28 from Human Oxytocin-neurophysin 1.
Host: Guinea Pig
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR