Anti-gamma MSH Guinea Pig Polyclonal Antibody

Proveedor: Biosensis
BSENGP-031-50EA 357 EUR
BSENGP-031-50
Anti-gamma MSH Guinea Pig Polyclonal Antibody
Anticuerpos
Proopiomelanocortin (POMC) is the precursor of the melanocortin peptides alpha, beta and gamma. POMC neurons in the arcuate nucleus of the hypothalamus function as key central regulators of food intake and homeostatic control of energy balance in both rodents and non-human primates.

Specificity was demonstrated by immunohistochemistry.
Application Information:
IHC. A concentration of 1 in 2000 is recommended for IHC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: gamma MSH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 77-88 from Human Proopiomelanocortin (POMC).
Host: Guinea Pig
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR