Anti-DJ-1 Mouse Monoclonal Antibody [clone: 4H4]

Proveedor: Biosensis
M-1573-100
BSENM-1573-100EA 302 EUR
BSENM-1573-100
Anti-DJ-1 Mouse Monoclonal Antibody [clone: 4H4]
Anticuerpos
Protein DJ-1 has many roles including protecting cells against oxidative stress and cell death (Ref: SwissProt). Mutations in the DJ-1 gene have been associated with rare forms of autosomal recessive early-onset Parkinson's disease.

The antibody reacts with a 21 kDa band by Western blot on whole HeLa cell lysate. It has also been used successfully for immunocytochemistry.
Application Information:
WB, IHC/IF. Suggested dilution of at least 1:1,000 for IHC/IF. Dilutions of 1:10,000 or lower is recommended for WB. This antibody reveals a prominent ~21 kDa band and stains mainly in cytoplasm of tissue culture cells. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: DJ-1
Clonality: Monoclonal
Clone: 4H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR