Anti-Saporin Rabbit Polyclonal Antibody
Proveedor: Biosensis
BSENR-025-100EA 302 EUR
BSENR-025-100
Anti-Saporin Rabbit Polyclonal Antibody
Anticuerpos
Saporin is a ribosome-inactivating protein (RIP) of type I. This monomeric RNA N-glycosidase purified from seeds of the plant Saponaria officinalis also known as Soapwort, is capable of specific depurination of eukaryotic ribosomes thus arresting protein synthesis. No ligand has been identified in saporin hence its inability to transverse the cell membrane. Due to its toxicity and stability of the structure, saporin has proven extremely useful for construction of immunotoxins. The expected molecular weight of the purified saporin is 29.5 kDa.
Confirmed to react with purified saporin.
Application Information:
IHC, Immunofluorescence, ELISA, Western Blot. A dilution of 1:200 to 1: 2000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Saporin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now