Anti-Semaphorin 6C Mouse Polyclonal Antibody

BSENMO-941-50EA 327 EUR
BSENMO-941-50
Anti-Semaphorin 6C Mouse Polyclonal Antibody
Semaphorin 6C is a transmembrane protein that shows growth cone collapsing activity on dorsal root ganglion neurons in vitro. It may be a stop signal for the dorsal root ganglion neurons, and possibly other neurons, in their target areas. It may also be involved in the maintenance and remodeling of neuronal connections.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Semaphorin 6C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Semaphorin 6C (27-126)
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR