The deduced 350-amino acid NPIP protein is membrane bound. Immunofluorescence microscopy demonstrated localization of NPIP to the nuclear membrane and colocalization with nuclear pore complex protein nucleoporin p62. NPIP is widely expressed and is thought to associate with the nuclear pore complex.
Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: NPIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Semaphorin 5A (393-499)
Host: Mouse
Isotype:
Reactivity: Human