Anti-Small EDRK-rich factor 1 Mouse Polyclonal Antibody

Proveedor: Biosensis
BSENMO-936-50EA 327 EUR
BSENMO-936-50
Anti-Small EDRK-rich factor 1 Mouse Polyclonal Antibody
Anticuerpos
There are 2 named isoforms, a long and a short isoform, produced by alternative splicing. The long isoform is predominantly expressed in heart, brain and skeletal muscle. Both isoforms are also expressed throughout the central nervous system including the spinal cord. Deletions of the EDRK-rich factor 1 gene, often accompany deletions of the neighboring SMN1 gene in spinal muscular atrophy (SMA) patients, and so it is thought that this gene may be a modifier of the SMA phenotype.

Specificity has been confirmed by direct ELISA against the antigen.
Application Information:
This antibody is recommended for ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Small EDRK-rich factor 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Small EDRK-rich factor 1 (1-82)
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR