Anti-NT3 Mouse Polyclonal Antibody

Proveedor: Biosensis
BSENMO-935-50EA 327 EUR
BSENMO-935-50
Anti-NT3 Mouse Polyclonal Antibody
Anticuerpos
NT3 is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It seems to promote the survival of visceral and proprioceptive sensory neurons. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse. NT3 is a secreted protein that is expressed in brain and peripheral tissues.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: NT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human NT3 (19-128)
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR