Anti-Protocadherin alpha-7 Mouse Polyclonal Antibody

Proveedor: Biosensis
BSENMO-948-50EA 327 EUR
BSENMO-948-50
Anti-Protocadherin alpha-7 Mouse Polyclonal Antibody
Anticuerpos
Protocadherin alpha-7 is 1 of 15 tandemly arranged genes within the PCDHA gene cluster. These 15 N-terminal 'variable' exons  are individually spliced to a downstream C-terminal constant region to form distinct PCDHA transcripts. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain.The PCDH proteins are cadherin-like cell surface proteins that are expressed in neurons and are present at synaptic junctions. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Protocadherin alpha-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human PCDH-alpha-7 (143-241)
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR