Anti-mouse TROY Rabbit Polyclonal Antibody

Proveedor: Biosensis
R-091-100
BSENR-091-100EA 302 EUR
BSENR-091-100
Anti-mouse TROY Rabbit Polyclonal Antibody
Anticuerpos
FUNCTION: Can mediate activation of c-Jun and NF-kappa-B. May promote caspase-independent cell death. Isoform 2 and isoform 3 may act as decoy receptors. SUBUNIT: Associates with TRAF1, TRAF2, TRAF3 and TRAF5. SUBCELLULAR LOCATION: Isoform 1, isoform 3, isoform 4: Cell membrane; single-pass type I membrane protein (Probable). Isoform 2: Secreted protein (Probable). ALTERNATIVE PRODUCTS: 4 named isoforms produced by alternative splicing. TISSUE SPECIFICITY: Highly expressed in adult brain, and in embryos from day 11-17, but not earlier. Detected in embryonic brain and epithelium, and at lower levels in adult heart, lung and liver. In neonatal mice, mainly in hair follicles and neuron-like cells in the cerebellum, but not in the skin epidermis. Isoform 3 was found in embryonic day 17.5 skin but not in brain and liver. SIMILARITY: Contains 3 TNFR-Cys repeats.

Specificity for TROY was confirmed by IHC.
Application Information:
IHC. Recommended to be used at a dilution of 1:500 to 1:2000 for immunohistochemistry. This antiserum has not yet been tested for western blot. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: mouse TROY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Mouse TROY protein (aa: 75-88)
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR