Anti-p75NTR Rabbit Polyclonal Antibody
Proveedor: Biosensis
BSENR-089-100EA 334 EUR
BSENR-089-100
Anti-p75NTR Rabbit Polyclonal Antibody
Anticuerpos
FUNCTION: Low affinity receptor which can bind to NGF, BDNF, NT-3, and NT-4. Can mediate cell survival as well as cell death of neural cells. SUBUNIT: Homodimer; disulfide-linked. Interacts with p75NTR-associated cell death executor. Interacts with TRAF2, TRAF4, TRAF6, PTPN13 and RANBP9. Interacts through TRAF6 with SQSTM1 which bridges NGFR to NTRK1. Interacts with BEX1 and NGFRAP1/BEX3. SUBCELLULAR LOCATION: Membrane; single-pass type I membrane protein. DOMAIN: Death domain is responsible for interaction with RANBP9. PTM: N- and O-glycosylated. PTM: O-linked glycans consist of Gal(1-3)GalNAc core elongated by 1 or 2 NeuNAc. PTM: Phosphorylated on serine residues. SIMILARITY: Contains 1 death domain. SIMILARITY: Contains 4 TNFR-Cys repeats.
IHC shows specific staining for p75NTR.
Application Information:
IHC, immunofluorescence. Recommended to be used at a dilution of 1:500 to 1:2000. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: p75NTR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: ECD Human p75
Host: Rabbit
Isotype: IgG
Reactivity: Rat