Anti-beta synuclein Rabbit Polyclonal Antibody

Proveedor: Biosensis
BSENR-056-100EA 345 EUR
BSENR-056-100
Anti-beta synuclein Rabbit Polyclonal Antibody
Anticuerpos
Beta-synuclein is a non-amyloid component of senile plaques found in Alzheimer disease. It could act as a regulator of SNCA aggregation. It protects nerurons from staurosporine and 6 hydroxy dopamine -stimulated capspase activation in a p53-dependent manner. It localises to the cytoplasm and it is predominantly expressed in the brain where it is most concentrated in presynaptic nerve terminals. This protein is phosphorylated. This protein is also associated with the disease Brain iron accumulation type 1 (NBIA1).

Less than 0.1% cross reactivity to human alpha synuclein
Application Information:
IHC, WB , immunoblot. A dilution of 1:500 to 1:2000 is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: beta synuclein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human beta synuclein protein (aa: 108-125)
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR