Anti-capsaicin receptor Rabbit Polyclonal Antibody

Proveedor: Biosensis
BSENR-076-100EA 302 EUR
BSENR-076-100
Anti-capsaicin receptor Rabbit Polyclonal Antibody
Anticuerpos
Responses evoked by low pH and heat, and capsaicin can be antagonized by capsazepine. SIMILARITY: Belongs to the transient receptor family. TrpV subfamily. SIMILARITY: Contains 3 ANK repeats.

Specificity was confirmed by IHC using frozen sections of rat dorsal root ganglia (DRG) and spinal cord. Due to sequence homology, similar staining is predicted in human DRG and spinal cord.
Application Information:
IHC. Use at 1:1000 to 1:2000 dilution. This antibody has not been tested in other applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: capsaicin receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human capsaicin receptor protein (aa: 608-621)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR