Anti-mouse ATPase Rabbit Polyclonal Antibody

Proveedor: Biosensis
BSENR-073-100EA 334 EUR
BSENR-073-100
Anti-mouse ATPase Rabbit Polyclonal Antibody
Anticuerpos
CATALYTIC ACTIVITY: ATP + H2O = ADP + phosphate. SUBCELLULAR LOCATION: Membrane; multi-pass membrane protein (By similarity). SIMILARITY: Belongs to the cation transport ATPase (P-type) family. Type V subfamily.

Immunohistochemistry shows specific staining for ATPase.
Application Information:
IHC. A dilution of 1:500 to 1:2000 is recommended for this application. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: mouse ATPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Mouse ATP13A2 protein (aa: 147-161)
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR