- Pagina principal
- Anticuerpos
- Anti-proBDNF Rabbit Polyclonal Antibody
Anti-proBDNF Rabbit Polyclonal Antibody
Proveedor: Biosensis
BSENR-087-100EA
302
EUR
BSENR-087-100
Anti-proBDNF Rabbit Polyclonal Antibody
Anticuerpos
BDNF belongs to the neurotrophin family and regulates the survival and differentiation of neurons during development. The alterations in BDNF expression induced by various kinds of brain insult including stress, ischemia, seizure activity and hypoglycemia, may contribute to some pathologies such as depression, epilepsy, Alzheimer's, and Parkinson's disease. Microglia release BDNF that may contribute to neuroinflammation and neuropathic pain. FUNCTION: Promotes the survival of neuronal populations that are all located either in the central nervous system or directly connected to it. Major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation of intrinsic neuronal excitability. SUBUNIT: Monomers and homodimers. Binds to NTRK2/TRKB. SUBCELLULAR LOCATION: Secreted protein. Post Translation Modification (PTM): The propeptide is N-glycosylated and glycosulfated. PTM: Converted into mature BDNF by plasmin (PLG) (By similarity). DISEASE: Defects in BDNF are a cause of congenital central hypoventilation syndrome (CCHS); also known as congenital failure of autonomic control or Ondine curse. CCHS is a rare disorder characterized by abnormal control of respiration in the absence of neuromuscular or lung disease, or an identifiable brain stem lesion. A deficiency in autonomic control of respiration results in inadequate or negligible ventilatory and arousal responses to hypercapnia and hypoxemia. CCHS is frequently complicated with neurocristopathies such as Hirschsprung disease that occurs in about 16% of CCHS cases. SIMILARITY: Belongs to the NGF-beta family.
Used in western blot, this antiserum detects a 35 kDa band corresponding to the molecular weight of proBDNF. No cross reactivity with other proneurotrophins was detected.
Application Information:
IHC, WB. A dilution of 1:1000 to 1:5000 is recommended for both applications. ICC: 1:500 to 1:2000, antibody works on 4% formaldehyde fixed cells. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
IHC, WB. A dilution of 1:1000 to 1:5000 is recommended for both applications. ICC: 1:500 to 1:2000, antibody works on 4% formaldehyde fixed cells. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: proBDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human BDNF precursor protein (aa: 69-82)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Antigen: proBDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human BDNF precursor protein (aa: 69-82)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Consulta de precio
Learn more
About VWR
Item(s) Added to Cart