- Pagina principal
- Anticuerpos
- Anti-MBP Sheep Polyclonal Antibody
Anti-MBP Sheep Polyclonal Antibody
Proveedor: Biosensis
BSENS-023-100EA
334
EUR
BSENS-023-100
Anti-MBP Sheep Polyclonal Antibody
Anticuerpos
Myelin is a membrane characteristic of the nervous tissue and functions as an insulator to increase the velocity of the stimuli being transmitted between a nerve cell body and its target. Myelin isolated from human and bovine nervous tissue is composed of approximately 80% lipid and 20% protein, and 30% of the protein fraction constitutes myelin basic protein (MBP). MBP is an 'intrinsically unstructured' protein with a high proportion (approximately 75%) of random coil, but postulated to have core elements of beta-sheet and alpha-helix. MBP is a major protein in CNS myelin and is expressed specifically in the nervous system. A detailed immunochemical examination of monoclonal and polyclonal antibody responses to MBP and its peptides has revealed the existence of as many as 27 antigenic determinants, many of them conformational. Topological mapping of the potential antigenic determinants onto a model of MBP secondary structure places these determinants within 11 separate regions of the molecule, including those portions that have been found to be encephalitogenic. The message for myelin basic protein is selectively translocated to the ends of the cell processes. Immunization with myelin-associated antigens including MBP significantly promotes recovery after spinal cord contusion injury in the rat model. FUNCTION: Is, with PLP, the most abundant protein component of the myelin membrane in the CNS. Has a role in both the formation and stabilization of this compact multilayer arrangement of bilayers. Each splice variant and charge isomer may have a specialized function in the assembly of an optimized, biochemically functional myelin membrane (By similarity). SUBUNIT: Homodimer (By similarity). SUBCELLULAR LOCATION: Myelin membrane; peripheral membrane protein; cytoplasmic side. Cytoplasmic side of myelin. TISSUE SPECIFICITY: Found in both the central and the peripheral nervous system. PTM: At least 5 charge isomers; C1 (the most cationic, least modified, and most abundant form), C2, C3, C4 and C5 (the least cationic form); are produced as a result of optional posttranslational modifications such as phosphorylation of serine or threonine residues, deamidation of glutamine or asparagine residues, citrullination and methylation of arginine residues. C1 and C2 are unphosphorylated, C3 and C4 are monophosphorylated and C5 is phosphorylated at two positions. SIMILARITY: Belongs to the myelin basic protein family.
This antiserum recognizes MBP in demyelinated nerve tissues. Immunohistochemical analysis of lesioned rat spinal cord indictaes a high level of specificity for this antiserum.
Application Information:
IHC. A dilution of 1:1000 to 1:4000 is recommended. Immunostaining for MBP of abnormal appearing oligodendrocytic process and cell bodies in demyelinating areas. This antibody recognises only areas of myelin degeneration when tested in injured spinal cord and lesioned sciatic nerves. It also stains discrete white matter in the brain of multiple system atrophy. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Application Information:
IHC. A dilution of 1:1000 to 1:4000 is recommended. Immunostaining for MBP of abnormal appearing oligodendrocytic process and cell bodies in demyelinating areas. This antibody recognises only areas of myelin degeneration when tested in injured spinal cord and lesioned sciatic nerves. It also stains discrete white matter in the brain of multiple system atrophy. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: aa: 68-86 of guinea pig MBP protein
Host: Sheep
Isotype: IgG
Reactivity: Rat, Human
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: aa: 68-86 of guinea pig MBP protein
Host: Sheep
Isotype: IgG
Reactivity: Rat, Human
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Consulta de precio
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Item(s) Added to Cart